missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAD1 Monoclonal Antibody (9B10)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA15829
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MAD1 Monoclonal Antibody for ICC/IF, Flow, IP
MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
Especificaciones
| MAD1 | |
| Monoclonal | |
| 0.9 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9Y6D9 | |
| MAD1L1 | |
| Recombinant full length Mad1 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
| 9B10 | |
| Unconjugated | |
| MAD1L1 | |
| AI173502; AW550425; coiled-coil domain-containing protein MAD1; hMAD1; HsMAD1; Mad1; MAD1 mitotic arrest deficient 1-like 1; MAD1 mitotic arrest deficient like 1; MAD1 mitotic arrest deficient-like 1; MAD1 mitotic arrest deficient-like 1 (yeast); MAD1L1; MAD1-like protein 1; mitotic arrest deficient 1-like 1; mitotic arrest deficient 1-like protein 1; Mitotic arrest deficient protein 1; Mitotic checkpoint MAD1 protein homolog; mitotic spindle assembly checkpoint protein MAD1; mitotic-arrest deficient 1, yeast, homolog-like 1; MXD1; PIG9; Spindle assembly checkpoint component MAD1; Tax-binding protein 181; TP53I9; tumor protein p53 inducible protein 9; TXBP181; YGL086W | |
| Mouse | |
| Protein A | |
| RUO | |
| 8379 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido