Descripción
The uvex 3, but even more robust.
- The MACSOLE™ S7 68633 boots with rubber outsole provides even stronger support for heavy-duty applications, making it particularly resistant to heat, chemicals and cuts.
- The MACSOLE™ rubber sole technology also impresses with its unbeatable slip resistance (SR), even in the cold.
- EN ISO 20345: S7
Especificaciones
Especificaciones
| Color | Black, Blue |
| Material | Rubber, Leather |
| Tamaño | 36 |
| Material del dedo del pie | Polymer |
| Desechables | Reusable |
| Sexo | Men's |
| Ajuste | Wide Width |
| Sin látex | No |
| Waterproof | Yes |
| Tipo de producto | Boots |