missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MA2 Polyclonal Antibody

Descripción
MA2 Polyclonal Antibody for IHC (P)
In patients with cancer, symptoms of limbic and brain-stem dysfunction may result from a para-neoplastic disorder. Two proteins, Ma1 and Ma2, were discovered by presence of autoantibodies against them. Antibodies to Ma1 and Ma2 proteins identify a paraneoplastic disorder that affects the limbic system, brain stem, and cerebellum. These autoantibodies were used to clone Ma2, which codes for a protein (Ma2) that was recognized by serum from the 10 patients, but not by serum from 344 control subjects. Ma2 was selectively expressed by normal brain tissue and by the testicular tumors of the patients. Ma2 shares homology with Ma1, a brain-testis-cancer gene related to other para-neoplastic syndromes and tumors.
Especificaciones
Especificaciones
| Antígeno | MA2 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 100 μg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.03% ProClin 300; pH 7 |
| génica | PNMA2 |
| N.º de referencia del gen | Q9UL42 |
| Alias de gen | 40 kDa neuronal protein; A830049P17Rik; KIAA0883; MA2; mKIAA0883; MM2; onconeuronal antigen MA2; Paraneoplastic antigen Ma2; paraneoplastic antigen Ma2 homolog; paraneoplastic Ma antigen 2; paraneoplastic neuronal antigen MM2; Pnma2; retrotransposon gag domain containing 2; RGAG2 |
| Símbolos de los genes | PNMA2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?