missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ M13 Phage coat protein Monoclonal Antibody (E1), Biotin

Descripción
MA1-34468 detects M13 from many samples.
M13 antibodies have numerous uses in screening phage display libraries, especially the screening of single chain antibody libraries. The display of repertoires of antibody fragments on the surface of filamentous phage offers a new way to produce immunoreagents with defined specificities. Phage derived antibody fragments offer a number of advantages over mouse monoclonal antibodies, such as better clearance from the blood, the possibility to select from human combinatorial libraries and the relative ease by which such fragments can be manipulated. The phage display technique thus facilitates the selection of antibody fragments of therapeutic value or research interest. Antibodies to M13 filamentous phage coat proteins are instrumental in the selection and detection of phages expressing specific antibody fragments or peptide sequences at their surface.
Especificaciones
Especificaciones
| Antígeno | M13 Phage coat protein |
| Aplicaciones | ELISA, Flow Cytometry, Western Blot |
| Clasificación | Monoclonal |
| Clon | E1 |
| Concentración | 1 mg/mL |
| Conjugado | Biotin |
| Formulación | PBS with 0.08% sodium azide |
| Alias de gen | Attachment protein G3P; G3P; Gene 3 protein; III; Minor coat protein |
| Especie del huésped | Mouse |
| Inmunógeno | Full length M13 phage coat protein; Exact epitope not determined. |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?