Learn More
Thermo Scientific™ GeneBLAzer™ M1 CHO-K1 DA Assay Kit
Código de producto. 10349243
Change view
Click to view available options
Cantidad:
1 placa
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
10349243 1 placa cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 10349243 Proveedor Thermo Scientific™ N.º de proveedor K1365
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
GeneBLAzer™ M1 CHO-K1 DA(Division Arrested) cells and M1-NFAT-bla CHO-K1 cells contain the human Acetylcholine (muscarinic) subtype 1 receptor (M1) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.
This cell line is available as dividing cells by requesting a quote for K1230 from dividingcells@invitrogen.com
GeneBLAzer™ M1 CHO-K1 DA(Division Arrested) cells and M1-NFAT-bla CHO-K1 cells contain the human Acetylcholine (muscarinic) subtype 1 receptor (M1) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.
DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both M1 CHO-K1 DA cells and M1-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of carbachol, (Figure 1). In addition, M1-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
The Musacarinic 1(M1) Receptor is localized principally in the cortical and hippocampal regions of the brain and has been implicated in various neurological conditions including depression, anxiety, and sleep disorders.
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso NM_000738
Entrada del ensayo Gen informador de beta lactamasa a base de células
Estado de célula Células de división detenida
Contenido y almacenamiento Almacenar en nitrógeno líquido inmediatamente después de su recepción, o descongelar para su uso inmediato
Método de detección Fluorescente
Para utilizar con (aplicación) GPCR
ID de gen (Entrez) 1128
Símbolo de gen M1
Incluye Células M1 CHO-K1 DA, kit de carga LiveBLAzer™-FRET B/G, 70 μg, solución D, 1 ml, 2 protocolos, certificado de análisis
Estado de la población Origen del clon unicelular
Línea de productos GeneBLAzer
Cantidad 1 placa
Gen marcador BLA (betalactamasa)
Entrada diana M1
Tipo Células de división detenida
Línea de células CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Mostrar más Mostrar menos
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.