Learn More
Melford Lisozima de clara de huevo de pollo
Código de producto. p-200086981
Change view
Click to view available options
Cantidad:
1 g
5 g
10 g
Tamaño de la unidad:
1 g
10 g
5 g
30018447 1 g 1 g
30018449 5 g 5 g
30018448 10 g 10 g
3 options
Código de producto. 30018447
Marca: Melford L381001.0
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
Purified from chicken egg white, 2x crystallized, dialyzed and supplied as a salt free lyophilized powder.
Dry powder stable for 3 - 5 years when stored dry at 2°C to 8°C.
Stock solution is stable up to 3 days at 4°C and up to 1 year at -20°C.
Especificaciones
CAS 12650-88-3
Cantidad 1 g