Learn More
Ly-6G Rat anti-Mouse, PE, Clone: 1A8, BD
Descripción
The 1A8 antibody reacts with Ly-6G, a 21-25kDa GPI-anchored protein. In the bone marrow, Ly-6G is expressed on the majority of the largest cells, which are predominantly granulocytes, and not on lymphoid or erythroid cells. In the periphery, it is expressed on granulocytes. The mAb RB6-8C5 (Cat. No. 557445/553123) recognizes both Ly-6G and Ly-6C and blocks the binding of mAb 1A8 to Ly-6G. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: 1A8
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Ly-6G-transfected EL4J cell line
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Especificaciones
Especificaciones
| Antígeno | Ly-6G |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 1A8 |
| Concentración | 0.2mg/mL |
| Conjugado | PE |
| Descripción | Ly6g; Lymphocyte antigen 6G; Lymphocyte antigen 6 complex, locus G; Gr1 |
| Formulación | Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. |
| Especie del huésped | Rat |
| Inmunógeno | Ly-6G-transfected EL4J cell line |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.