Learn More
Ly-6G and Ly-6C Rat anti-Mouse, Unlabeled, Clone: RB6-8C5, BD
Rat Monoclonal Antibody
216.40€ - 319.63€
Especificaciones
| Antígeno | Ly-6G and Ly-6C |
|---|---|
| Clon | RB6-8C5 |
| Concentración | 0.5mg/mL |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
15887008
|
BD Biosciences
553123 |
0.5 mg |
320.00€ 319.63€ / cada uno Ahorro 0.37€ 0.12% Descuento |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
10269334
|
BD Biosciences
550291 |
1 mL |
217.00€ 216.40€ / cada uno Ahorro 0.60€ 0.28% Descuento |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
The RB6-8C5 antibody reacts with a common epitope on Ly-6G and Ly-6C, previously known as the myeloid differentiation antigen Gr-1. In the bone marrow, the level of antigen expression is directly correlated with granulocyte differentiation and maturation. The antigen is also expressed on the monocyte lineage in the bone marrow, but not on erythroid cells. In the periphery, RB6-8C5 antibody recognizes granulocytes (neutrophils and eosinophils) and monocytes. The RB6-8C5 mAb is a component of the “lineage cocktail” used in studies of hematopoietic lineages. The mAb 1A8 (Cat. No. 551461) specifically recognizes Ly-6G, but not Ly-6C. This antibody is routinely tested by immunohistochemistry. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: RB6-8C5
Isotype: IgG2b κ
Species Reactivity: Mouse
Immunohistochemistry, Immunohistochemistry (Frozen)
Especificaciones
| Ly-6G and Ly-6C | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Immunology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2b κ | |
| Affinity Purified | |
| Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1 |
| RB6-8C5 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Mouse | |
| Not Reported | |
| Primary | |
| Store undiluted at 4°C |
For Research Use Only.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.