missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-49I Monoclonal Antibody (YLI-90)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA528668
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ly-49I Monoclonal Antibody for Flow
Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
Especificaciones
| Ly-49I | |
| Monoclonal | |
| 0.5 mg/mL | |
| BBS with no preservative; pH 8.2 | |
| 0 | |
| Klra9 | |
| CHO-K1 cells transfected with B6 allele of Ly49I gene. | |
| 1 mL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| YLI-90 | |
| Unconjugated | |
| Klra9 | |
| 5E6; Killer cell lectin-like receptor 3; killer cell lectin-like receptor subfamily A, member 9; Klra3; Klra9; Ly-49; Ly49C; Ly-49c; Ly49I; Ly49I receptor; Ly-49I receptor; LY49I1; Lymphocyte antigen 49c; Nk2.1; T-cell surface glycoprotein Ly-49C | |
| Mouse | |
| Purified | |
| RUO | |
| 16640 | |
| 4°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido