Learn More
LXR alpha/NR1H3 Antibody (S07-6A4), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00€ - 645.00€
Especificaciones
Antígeno LXR alpha/NR1H3
Clon S07-6A4
Concentración 0.3 mg/mL
Dilución Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
18626323
Ver documentos
Novus Biologicals
NBP3-19682-100ul
100 μg
645.00€
100 microlitros
Ver productos alternativos
Ahorre hasta 
»
18624943
Ver documentos
Novus Biologicals
NBP3-19682-50ul
50 μg
421.00€
50 microlitros
Ver productos alternativos
Ahorre hasta 
»
Descripción
Descripción
LXR alpha/NR1H3 Monoclonal antibody specifically detects LXR alpha/NR1H3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
LXR alpha/NR1H3
0.3 mg/mL
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
10062
IgG
Affinity purified
S07-6A4
Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse, Rat
alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1
A synthetic peptide of human LXR alpha/NR1H3 (Uniprot # Q13133)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.