missing translation for 'onlineSavingsMsg'
Learn More

LX-1031, MedChemExpress

Código de producto. 16441687
Encompass_Preferred
Change view
Click to view available options
Cantidad:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
2 miligramos
25 miligramos
5 miligramos
50 miligramos
Código de producto. Cantidad unitSize
16441687 50 mg 50 miligramos
16493346 25 mg 25 miligramos
16486086 100 mg 100 miligramos
16485508 10 mg 10 miligramos
16428227 5 mg 5 miligramos
16400196 2 mg 2 miligramos
6 options
This item is not returnable. View return policy

Product Code. 16441687

Brand: Medchem Express HY13041

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress LX-1031 is a potent, orally available tryptophan 5-hydroxylase (TPH) inhibitor that reduces serotonin (5-HT) synthesis peripherally.

LX-1031 is a potent, orally available tryptophan 5-hydroxylase (TPH) inhibitor that reduces serotonin (5-HT) synthesis peripherally.

Specifications

Nombre del producto químico o material LX-1031
CAS 945976-76-1
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C28H25F3N4O4
Cantidad 50 mg
Información de solubilidad DMSO : ≥ 34 mg/mL (63.14 mM)
SMILES NC1=NC(C2=CC=C(C=C2)C[C@@H](C(O)=O)N)=CC(O[C@H](C3=CC=C(C4=CC(OC)=CC=C4)C=C3)C(F)(F)F)=N1
Peso molecular (g/mol) 538.52
Formula Weight (peso de la fórmula) 538.52
Porcentaje de pureza 98.32%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.