Learn More
Value Plastics Luer Adapter Fittings, Tee, Polypropylene
Tienda Cole-Parmer Productos
Click to view available options
Anchura (imperial):
29/32 in
37/64 in
Tamaño de la unidad:
paquete de 25
Descripción
Quick and easy assembly and disassembly without the need for tools. Luer fittings allow for a simple yet secure connection. Conforms to ISO-594-1 and ISO 594-2 standards. Constructed of ADCF polypropylene
Especificaciones
Especificaciones
| Certificaciones/conformidad | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Cantidad de embalaje | 25/Pk. |
| Conexión | Female Luer Thread, Female Luer Thread |
| Anchura (imperial) | 37/64 in |