missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRFIP1 Polyclonal Antibody

Descripción
A suggested positive control is human colon tissue lysate. PA5-21114 can be used with blocking peptide PEP-1228.
Leucine-rich repeat FLI-I interacting protein 1 (LRRFIP1/FLAP-1) is a MyD88-interacting signal protein which mediates either induction or repression of TLR signaling. Flap-1 directly interacts with β-catenin and activates β-catenin-dependent transcription activity. Flap-1 or LRRFIP1 was also shown to bind the TNF- -308 promoter polymorphism site, suggesting its possible role in the regulation of TNF- production. FLAP-1 along with LRRFIP2 and FLIIH serve as a balance in coordinating inflammatory response after infection, and may act as a novel target in the control of agonist-induced TLR-mediated inflammatory and autoimmune diseases.
Especificaciones
Especificaciones
| Antígeno | LRRFIP1 |
| Aplicaciones | Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% sodium azide |
| génica | LRRFIP1 |
| N.º de referencia del gen | Q32MZ4 |
| Alias de gen | AU024550; Flap; FLAP (FLI LRR associated protein); FLAP1; flap-1; Fliiap1; FLI-LRR-associated protein 1; GC-binding factor 2; GCF2; GCF-2; H186 FLAP; HUFI1; HUFI-1; leucine rich repeat (in FLII) interacting protein 1; leucine-rich repeat flightless-interacting protein 1; LRR binding FLII interacting protein 1; LRR FLII-interacting protein 1; Lrrfip1; NEDD8-conjugating enzyme; TAR RNA-interacting protein; TRIP |
| Símbolos de los genes | LRRFIP1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?