missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC50 Monoclonal Antibody (OTI3F9), TrueMAB™, OriGene
Description
The protein encoded by this gene is cilium-specific and is required for the stability of the ciliary architecture. It is involved in the regulation of microtubule-based cilia and actin-based brush border microvilli. Mutations in this gene are associated with primary ciliary dyskinesia-13.
Specifications
Specifications
| Antígeno | LRRC50 |
| Aplicaciones | Flow Cytometry, Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI3F9 |
| Concentración | 0.76 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | DNAAF1 |
| N.º de referencia del gen | Q6AYH9, Q8NEP3, Q9D2H9 |
| Alias de gen | CILD13, LRRC50, ODA7, swt |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?