missing translation for 'onlineSavingsMsg'
Learn More

LPIN1 Monoclonal Antibody (OTI2C8), TrueMAB™, OriGene

Product Code. 17546353
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Unit Size:
100 microlitros
30 microlitros
Product Code. Cantidad unitSize
17546353 30 μL 30 microlitros
17526353 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 17546353

Brand: Origene Technologies TA806239S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LPIN1 Monoclonal antibody specifically detects LPIN1 in Human samples. It is validated for Western Blot

Specifications

Antígeno LPIN1
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI2C8
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica SPP1
N.º de referencia del gen P10451
Alias de gen 4631420P06; DKFZp781P1796; Fatty liver dystrophy protein; Fld; KIAA0188; lipin 1; Lipin1; lipin-1; LPIN1; mKIAA0188; PAP1; Phosphatidate phosphatase LPIN1
Especie del huésped Mouse
Inmunógeno Human recombinant protein fragment corresponding to amino acids 17-314 of human SPP1 produced in E.coli.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6696
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.