Learn More
Low-Level Rare Earth Carbonatite, RM, ARMI∣MBH
Código de producto. 30319138
Change view
Click to view available options
Cantidad:
90 g
Tamaño de la unidad:
90 g
30319138 90 g 90 g
1 options
Código de producto. 30319138
Marca: ARMI/MBH MBHREELO22P
ARMI/MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. Their offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
TRUSTED_SUSTAINABILITY
Especificaciones
Nombre del producto químico o material Aluminium∣Aluminium oxide∣Barium∣Barium oxide∣Calcium∣Calcium oxide∣Cerium∣Cerium(IV) oxide∣Dysprosium∣Europium∣Holmium∣Iron∣Iron (III) Oxide∣Lanthanum∣Lanthanum(III) Oxide∣Lead∣Loss on ignition∣Lutetium∣Magnesium∣Magnesium oxide∣Manganese∣Manganese(II) oxide∣Neodymium∣Neodymium(III) oxide∣Niobium∣Phosphorus∣Phosphorus pentoxide∣Potassium∣Potassium oxide∣Praseodymium∣Praseodymium oxide∣Rubidium∣Samarium∣Scandium∣Silicon∣Silicon dioxide∣Strontium∣Strontium oxide∣Sulfur∣Sulfur trioxide∣Thorium∣Titanium∣Titanium dioxide∣Total Light Rare Earth Element Oxides∣Total Light Rare Earth Elements∣Uranium∣Ytterbium∣Yttrium
Cantidad 90 g
Peso 90 g
Formulario Powder
Concentration or Composition (by Analyte or Components) Titanium 0.012 %∣Praseodymium 0.013 %∣Praseodymium oxide 0.015 %∣Titanium dioxide 0.019 %∣Manganese 0.028 %∣Potassium 0.029 %∣Neodymium 0.034 %∣Potassium oxide 0.035 %∣Manganese(II) oxide 0.037 %∣Neodymium(III) oxide 0.04 %∣Phosphorus 0.041 %∣Lutetium 0.07 mg/kg∣Phosphorus pentoxide 0.095 %∣Lanthanum 0.1 %∣Lanthanum(III) Oxide 0.12 %∣Cerium 0.14 %∣Aluminium 0.16 %∣Cerium(IV) oxide 0.18 %∣Strontium 0.19 %∣Iron 0.2 %∣Sulfur 0.2 %∣Strontium oxide 0.22 %∣Iron (III) Oxide 0.29 %∣Total Light Rare Earth Elements 0.29 %∣Aluminium oxide 0.31 %∣Total Light Rare Earth Element Oxides 0.35 %∣Holmium 0.38 mg/kg∣Sulfur trioxide 0.49 %∣Ytterbium 0.5 mg/kg∣Barium 0.67 %∣Barium oxide 0.75 %∣Scandium 0.85 mg/kg∣Silicon 0.88 %∣Rubidium 1.5 mg/kg∣Magnesium 1.7 %∣Uranium 1.8 mg/kg∣Silicon dioxide 1.9 %∣Niobium 2.5 mg/kg∣Dysprosium 2.6 mg/kg∣Magnesium oxide 2.8 %∣Europium 4.2 mg/kg∣Thorium 9.8 mg/kg∣Yttrium 10 mg/kg∣Samarium 24 mg/kg∣Calcium 35.94 %∣Loss on ignition 42.1 %∣Calcium oxide 50.3 %∣Lead 149 mg/kg