missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LONP1 Polyclonal Antibody
Descripción
Immunogen sequence: PAEVLMVEVE NVVHEDFQVT EEVKALTAEI VKTIRDIIAL NPLYRESVLQ MMQAGQRVVD NPIYLSDMGA ALTGAESHEL QDVLEETNIP KRLYKALSLL KKEFELSKLQ QRLGREVEEK IKQTHRKYLL QEQLKIIKKE LGLEKDDKDA Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.
This gene encodes a mitochondrial matrix protein that belongs to the Lon family of ATP-dependent proteases. This protein mediates the selective degradation of misfolded, unassembled or oxidatively damaged polypeptides in the mitochondrial matrix. It may also have a chaperone function in the assembly of inner membrane protein complexes, and participate in the regulation of mitochondrial gene expression and maintenance of the integrity of the mitochondrial genome. Decreased expression of this gene has been noted in a patient with hereditary spastic paraplegia. Alternatively spliced transcript variants have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | LONP1 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.05 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | LONP1 |
| N.º de referencia del gen | P36776 |
| Alias de gen | 1200017E13Rik; CODASS; h LonHS; HAMAP-Rule:MF_03120}; hLON; hLON ATP-dependent protease; Lon; lon peptidase 1, mitochondrial; lon protease homolog, mitochondrial; lon protease homolog, mitochondrial {ECO:0000255; Lon protease-like protein; lon protease-like protein {ECO:0000255; LONHs; LONP; LONP {ECO:0000255; LONP1; Mitochondrial ATP-dependent protease Lon; mitochondrial ATP-dependent protease Lon {ECO:0000255; mitochondrial lon protease-like protein; PIM1; protease, serine, 15; Prss15; Serine protease 15; serine protease 15 {ECO:0000255 |
| Símbolos de los genes | LONP1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?