missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lodoxamide, MedChemExpress
Description
Lodoxamide (U-42585E free acid) is an antiallergic compound acting as a mast-cell stabilizer for the treatment of asthma and allergic conjunctivitis.
Specifications
Specifications
| Nombre del producto químico o material | Lodoxamide |
| Forma física | Solid |
| CAS | 53882-12-5 |
| Color | Amarillo claro |
| Cantidad | 200 mg |
| Notas de grado de pureza | Research |
| Fórmula molecular | C11H6ClN3O6 |
| Para utilizar con (aplicación) | COVID-19-immunoregulation |
| Sinónimo | U-42585E free acid |
| Información de solubilidad | DMSO : 50 mg/mL (160.45 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?