Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points Traceable Calibration
Código de producto. 17863068
Click to view available options
Intervalo de temperatura (métrico):
-10°C a +50°C
De -10 ° a +360 °C
De -10 °C a +110 °C
De -10 °C a +210 °C
De -10 °C a +260 °C
De -10 °C a +300 °C
De -10 °C a +330 °C
De -1 °C a 101 °C
De -1 °C a 51 °C
De -20 °C a +110 °C
De -20 °C a +150 °C
De -35 °C a +50 °C
Subdivisión:
0,1 °C
0,2 °C
0,5 °C
1 °C
2 °C
Tipo de inmersión:
inmersión parcial
inmersión total
Longitud (métrico):
15,5 cm
30,5 cm
40,5 cm
missing translation for 'unitSize'
cada uno
This item is not returnable. View return policy
Product Code. 17863068
missing translation for 'mfr': Brannan™ 44/821/0 + 49/101/0
This item is not returnable. View return policy
LO-tox™ lab thermometer complete with calibration certificate.
  • LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications
Puntos de calibración 0°C, 20°C, 35°C
Certificaciones/conformidad UK Traceable to National Standards ISO 9001
Tipo de inmersión inmersión total
Longitud (métrico) 40,5 cm
Material Glass
Relleno del termómetro Alcohol azul
Temperatura máx. (imperial) 124°F
Temperatura máxima +51°C
Temperatura mín. (imperial) 30°F
Temperatura mín. (métrico) -1°C
Intervalo de temperatura (imperial) 30°F to 124°F
Intervalo de temperatura (métrico) De -1 °C a 51 °C
Exactitud ± 0.5°C at 35°C
Subdivisión 0,1 °C
Anchura (métrico) 1.2 cm
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.