missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMP7/PSMB8 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Marca: Novus Biologicals NBP1-51957
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LMP7/PSMB8 Polyclonal specifically detects LMP7/PSMB8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Especificaciones
| LMP7/PSMB8 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| beta5i, D6S216E, EC 3.4.25.1, LMP7MGC1491, Low molecular mass protein 7, low molecular weight protein 7, Macropain subunit C13, Multicatalytic endopeptidase complex subunit C13, protease component C13, proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctionalpeptidase 7), proteasome catalytic subunit 3i, Proteasome component C13, proteasome subunit beta 5i, proteasome subunit beta type-8, Proteasome subunit beta-5i, proteasome subunit Y2, proteasome-related gene 7, PSMB5iD6S216, Really interesting new gene 10 protein, RING10proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctionalprotease 7), Y2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_004150.1; NP_683720.2). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.01 - 0.03 ug/ml, Immunohistochemistry 3.75 ug/ml, Immunohistochemistry-Paraffin 3.75 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| NP_004150.1 | |
| PSMB8 | |
| Peptide with sequence C-DVSDLLHQYREANQ corresponding to C-Terminus according to NP_683720.2. | |
| 0.1 mg | |
| Stem Cell Markers | |
| 5696 | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido