Learn More
Invitrogen™ LMBR1L Polyclonal Antibody
Código de producto. 13241089
Change view
Click to view available options
Cantidad:
400 μL
Tamaño de la unidad:
100 microgramos
13241089 400 μL 100 microgramos
1 options
Código de producto. 13241089 Proveedor Invitrogen™ N.º de proveedor PA526795
Rabbit Polyclonal Antibody
LMBR1L Polyclonal Antibody for Western Blot, IHC (P), Flow
LMBR1L, also known as LIMR (lipocalin-1-interacting membrane receptor), interacts with lipocalin-1 (Lcn-1), a lipocalin member produced by a number of secretory glands and tissues and known to bind a variety of lipophilic compounds. LMBR1L is a 489-amino acid protein with a predicted molecular mass of 55 kDa and 9 putative transmembrane helices. LMBR1L has been shown to mediate cellular uptake of LMBR1L, thus it is characterized as a endocytic receptor. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno LMBR1L
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.09% sodium azide; pH 7.4
génica LMBR1L
N.º de referencia del gen Q6UX01
Alias de gen 1110013E13Rik; AA415653; D15Ertd735e; KIAA1174; limb development membrane protein 1 like; limb development membrane protein 1-like; limb region 1 homolog-like; limb region 1 like; limb region 1 protein homolog-like; limb region 1-like homolog; limb region 1-like protein -like; LIMR; lipocalin-1 interacting membrane receptor; lipocalin-1-interacting membrane receptor; lipocalin-interacting membrane receptor; Lmbr1l; Protein LMBR1L; RGD1309631; UNQ458/PRO783; Uteroglobin receptor
Símbolos de los genes LMBR1L
Especie del huésped Rabbit
Inmunógeno KLH conjugated synthetic peptide between 462-489 amino acids from the C-terminal region of human LMBR1L.
Método de purificación Antigen Affinity Chromatography, Protein A
Cantidad 400 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 55716
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG