Learn More
Agilent BioTek Liquid Handling Control Software, USB Delivery
Tienda Agilent BioTek Productos
Click to view available options
Para utilizar con (equipo):
Lavadores de microplacas, dispensadores y sistemas de manipulación de líquidos Agilent BioTek
Tamaño de la unidad:
cada uno
Descripción
- LHC software makes it easy to program, even the most complex protocols with a choice of deployment
- Protocols can be run directly from the computer or downloaded to the instrument for convenient onboard recall and operation
- Full instrument control
- For use with 405 TS Washer, 405 LS Washer, ELx405 Select Deep Well Washer, 50 TS Washer, MultiFlo Dispenser, MultiFlo FX Dispenser, EL406 Washer Dispenser, 406 FX Washer Dispenser
- Batch processing with connected BioStack Microplate Stacker or BioSpa 8 Automated Incubator
- Seamless robotic integration to commercially available robotic systems
Especificaciones
Especificaciones
| Incluye | USB Flash Drive Delivery |
| Para utilizar con (equipo) | Lavadores de microplacas, dispensadores y sistemas de manipulación de líquidos Agilent BioTek |
| Tipo | Liquid Handling Control Software |