missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIPG Monoclonal Antibody (OTI7D1), TrueMAB™, OriGene
Description
LIPG Monoclonal antibody specifically detects LIPG in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Specifications
Specifications
| Antígeno | LIPG |
| Aplicaciones | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI7D1 |
| Concentración | 0.97 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | TLDC1 |
| N.º de referencia del gen | Q6P9B6 |
| Alias de gen | 3110013K01Rik; EDL; EL; endothelial cell-derived lipase; Endothelial lipase; endothelial-derived lipase; lipase; lipase G; lipase G, endothelial type; lipase, endothelial; LIPG; lipoprotein lipase H; lipose, endothelial; mEDL; PL; PRO719; UNQ387/PRO719 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?