missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIN28A Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15920935
Change view
Click to view available options
Cantidad:
200 μL
Tamaño de la unidad:
200 microlitros
Código de producto. Cantidad unitSize
15920935 200 μL 200 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15920935

Marca: Invitrogen™ PA572262

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

LIN28A Polyclonal Antibody for Western Blot

Lin28 is a transcription factor that was first identified through its key role in the pathway of developmental timing in C. elegans. The role of Lin28 in development suggested that it might be useful in the creation of stem cells that might be beneficial in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing Lin28 in addition to the transcription factors POU5F1, Sox2, and NANOG in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Lin28, Sox2, POU5F1 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno LIN28A
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.45 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.09% sodium azide; pH 7.4
génica LIN28A
N.º de referencia del gen Q803L0
Alias de gen AL024421; CSDD1; ENSMUSG00000070700; FLJ12457; Gm10299; Lin28; LIN-28; lin-28 homolog A; lin-28 homolog A (C. elegans); Lin28a; lin-28A; Protein lin-28 homolog A; RGD1566408; RNA-binding protein LIN-28; sb:cb650; si:ch211-232d9.4; similar to RNA-binding protein LIN-28; testis expressed gene 17; testis-expressed protein 17; Tex17; ZCCHC1; zgc:55584; Zinc finger CCHC domain-containing protein 1; zinc finger, CCHC domain containing 1
Símbolos de los genes LIN28A
Especie del huésped Rabbit
Inmunógeno KLH conjugated synthetic peptide between 85-119 amino acids from the Central region of DANRE lin28a.
Método de purificación Antigen Affinity Chromatography, Protein A
Cantidad 200 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 394066
Especies diana Zebrafish
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.