Learn More
LIMK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16810024 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16810024 20 μL 20 microlitros
16800024 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16810024 Proveedor Proteintech N.º de proveedor 123501AP20UL
Rabbit Polyclonal Antibody
Proteins containing LIM motifs are typically involved in cell fate determination and growth control. A family of proteins designated LIM kinases, including LIMK-1 and LIMK-2, has been identified. LIMK-1 has been shown to regulate the stabilization of F-Actin structures and cofilin activity, indicating that LIMK-1 plays a role in a signaling pathway involved in the regulation of cell motility and morphogenesis. LIMK-1 inhibits neuronal differentiation of PC12 cells, and is thought to act by interfering with events downstream of MAPK activation. Expression patterns of LIMK-1 and LIMK-2 suggest that these proteins may have different functions during development. A truncated form of LIMK-2 has been identified in adult testis that is thought to arise from an alternative initiation exon.
Especificaciones
Antígeno LIMK2
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.25 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica LIMK2
N.º de referencia del gen O54785, P53670, P53671
Alias de gen LIM domain kinase 2, LIMK 2, LIMK2
Símbolos de los genes LIMK2
Especie del huésped Rabbit
Inmunógeno LIMK2 Fusion Protein Ag3011
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16886, 29524, 3985
Especies diana Mouse, Rat, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG