Learn More
LIGHT/TNFSF14 Antibody (906909), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals FAB17943H
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LIGHT/TNFSF14 Monoclonal antibody specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Especificaciones
| LIGHT/TNFSF14 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| 0.1 mL | |
| Apoptosis | |
| 8740 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 906909 | |
| Flow Cytometry, CyTOF-ready | |
| CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
| Mouse myeloma cell line NS0-derived recombinant mouse LIGHT/TNFSF14, Asp72-Val239, Accession # Q9QYH9 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG1 |