missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX2 Monoclonal Antibody (OTI3D3), TrueMAB™, OriGene
Description
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.
Specifications
Specifications
| Antígeno | LHX2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI3D3 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | LHX2 |
| N.º de referencia del gen | P50458 |
| Alias de gen | LH2, hLhx2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?