missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGALS3BP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23307-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.Especificaciones
| LGALS3BP | |
| Polyclonal | |
| Unconjugated | |
| LGALS3BP | |
| 90K, BTBD17B, Galectin 3 binding protein, LGALS3BP, M2BP, Mac 2 binding protein, MAC 2 BP, MAC2BP, Tumor associated antigen 90K | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19039, 3959 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q07797, Q08380 | |
| Lgals3bp | |
| LGALS3BP Fusion Protein Ag19700 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido