missing translation for 'onlineSavingsMsg'
Learn More

LGALS3BP Monoclonal Antibody (OTI1F8), TrueMAB™, OriGene

Product Code. p-200012972
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Unit Size:
100 microlitros
30 microlitros
Product Code. Cantidad unitSize
17569321 30 μL 30 microlitros
17549321 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 17569321

Brand: Origene Technologies TA503454S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo requiere una actualización de precios antes de ser solicitado.

Solicite el precio de este artículo aquí.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LGALS3BP Monoclonal antibody specifically detects LGALS3BP in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antígeno LGALS3BP
Aplicaciones Flow Cytometry, Immunocytochemistry, Western Blot
Clasificación Monoclonal
Clon OTI1F8
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica LGALS3BP
N.º de referencia del gen Q08380
Alias de gen 90K; basement membrane autoantigen p105; BTBD17B; CyCAP; Cyp-C-associated protein; galectin 3 binding protein; galectin-3-binding protein; gp90; L3 antigen; Lectin; lectin galactoside-binding soluble 3 binding protein; lectin galactoside-binding soluble 3-binding protein; lectin, galactoside binding soluble 3 binding protein; lectin, galactoside-binding, soluble, 3 binding protein; Lgals3bp; M2BP; mac-2 BP; mac-2-binding protein; MAC2BP; MAC-2BP; MAC-2-BP; mama; peptidylprolyl isomerase C-associated protein; Ppicap; Protein 90K; Protein MAMA; serum protein 90K; similar to Galectin-3 binding protein precursor (Lectin galactoside-binding soluble 3 binding protein) (Mac-2 binding protein) (Mac-2 BP) (MAC2BP) (Tumor-associated antigen 90K); TANGO10B; transport and golgi organization 10 homolog B; tumor-associated antigen 90K
Especie del huésped Mouse
Inmunógeno Human recombinant protein fragment corresponding to amino acids 19-300 of human LGALS3BP produced in E.coli.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3959
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.