Learn More
Invitrogen™ Leptin Receptor Polyclonal Antibody
Descripción
PA1-053 detects leptin (OB-R) from human, mouse, and rat tissues. PA1-053 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~100 kDa and ~125 kDa protein representing the short and long forms of OB-R from mouse brain extract. This antibody also detects two smaller, unrelated proteins of ~31 kDa and 35 kDa which are unable to be competed with the peptide in mouse liver extract. The PA1-053 immunogen is a synthetic peptide corresponding to residues K(577) E I Q W K T H E V F D A K S K S A(594) of the extracellular domain of rat OB-R. This sequence is completely conserved in mouse OB-R. PA1-053 immunizing peptide (Cat. # PEP-014) is available for use in neutralization and control experiments.
Especificaciones
Especificaciones
| Antígeno | Leptin Receptor |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 10 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% sodium azide |
| génica | Lepr |
| N.º de referencia del gen | P48356, P48357, Q62959 |
| Alias de gen | B219; CD antigen CD295; CD295; CD295 antigen; db; diabetes; Fa; HuB219; HuB219.2; LEPR; LEP-R; Leprb; LEPRD; LEPROT; leptin receptor; Leptin receptor (fatty); leptin receptor gene-related protein; Modb1; OB receptor; obese-like; obl; Obr; OB-R; OB-RGRP |
| Símbolos de los genes | Lepr |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.