missing translation for 'onlineSavingsMsg'
Learn More

PeproTech Leptin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Código de producto. p-7245180
Change view
Click to view available options
Cantidad:
2 x 500 μg
25 μg
50 μg
500 μg
Tamaño de la unidad:
1 miligramo
25 microgramos
50 microgramos
500 microgramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17835833 25 μg 25 microgramos
17855013 50 μg 50 microgramos
17805683 2 x 500 μg 1 miligramo
30054480 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17835833 Proveedor PeproTech N.º de proveedor 500P185GBT25UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody

AA Sequence of recombinant protein: MVPIHKVQDD TKTLIKTIVT RINDISHTQS VSARQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQILTSLP SQNVLQIAHD LENLRDLLHL LAFSKSCSLP QTRGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDLSPEC. Preparation: Produced from sera of goats immunized with highly pure Recombinant Rat Leptin. Anti-Rat Leptin-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat Leptin by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat Leptin (500-P185G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat Leptin. Western Blot: To detect Rat Leptin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat Leptin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P185GBT-1MG will be provided as 2 x 500 μg

Leptin is a 167 amino acid long protein hormone with important effects in regulating body weight, metabolism and reproductive function. Leptin is approximately 16 kDa in mass and encoded by the obese (ob) gene. Leptin is expressed predominantly by adipocytes, which fits with the idea that body weight is sensed as the total mass of fat in the body. Smaller amounts of leptin are also secreted by cells in the epithelium of the stomach and in the placenta. Leptin receptors are highly expressed in areas of the hypothalamus known to be important in regulating body weight, as well as in T lymphocytes and vascular endothelial cells. Leptin's effects on body weight are mediated through effects on hypothalamic centers that control feeding behavior and hunger, body temperature and energy expenditure. Leptin is involved in regulating food intake, energy expenditure, and adiposity through hypothalamic leptin receptors. Leptin promotes hematopoiesis, angiogenesis, wound healing, inflammation, immune responses, influences pubertal development and fetal growth. Studies have investigated the role of leptin in obesity, anorexia nervosa, insulin resistance, and hypertension. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans can be caused by defects in leptin production.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Leptin
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Biotin
Formulación PBS with no preservative
génica Lep
N.º de referencia del gen P50596
Alias de gen CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin
Símbolos de los genes Lep
Especie del huésped Goat
Inmunógeno E.coli-derived Recombinant Rat Leptin.
Método de purificación Antigen affinity chromatography
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 25608
Especies diana Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.