missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Leptin Monoclonal Antibody (3)

Código de producto. 11514132
Change view
Click to view available options
Cantidad:
400 μL
Tamaño de la unidad:
200 microlitros
Código de producto. Cantidad unitSize
11514132 400 μL 200 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11514132

Marca: Invitrogen™ ABS0220302

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

ABS 022-03-02 detects Leptin in human samples. ABS 022-03-02 has been successfully used in ELISA and immunohistochemistry procedures. ABS 022-03-02 binds free Leptin in solution. In sandwich ELISA, ABS 022-03-02 works as a capture antibody with ABS 022-11-02 as a detection antibody. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

Leptin is a 167 amino acid long protein hormone with important effects in regulating body weight, metabolism and reproductive function. Leptin is approximately 16 kDa in mass and encoded by the obese (ob) gene. Leptin is expressed predominantly by adipocytes, which fits with the idea that body weight is sensed as the total mass of fat in the body. Smaller amounts of leptin are also secreted by cells in the epithelium of the stomach and in the placenta. Leptin receptors are highly expressed in areas of the hypothalamus known to be important in regulating body weight, as well as in T lymphocytes and vascular endothelial cells. Leptin's effects on body weight are mediated through effects on hypothalamic centers that control feeding behavior and hunger, body temperature and energy expenditure. Leptin is involved in regulating food intake, energy expenditure, and adiposity through hypothalamic leptin receptors. Leptin promotes hematopoiesis, angiogenesis, wound healing, inflammation, immune responses, influences pubertal development and fetal growth. Studies have investigated the role of leptin in obesity, anorexia nervosa, insulin resistance, and hypertension. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans can be caused by defects in leptin production.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Leptin
Aplicaciones ELISA, Immunohistochemistry
Clasificación Monoclonal
Clon 3
Concentración 1.09 mg/mL
Conjugado Unconjugated
Formulación PBS with 15mM sodium azide; pH 7.4
génica Lep
N.º de referencia del gen P41159
Alias de gen CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin
Símbolos de los genes Lep
Especie del huésped Mouse
Inmunógeno Purified, recombinant Leptin.
Método de purificación Protein A
Cantidad 400 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3952
Especies diana Human
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.