missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LEPRE1 Monoclonal Antibody (OTI1H3)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA526043
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LEPRE1 Monoclonal Antibody for Western Blot, IHC (P), Flow
Basement membrane-associated chondroitin sulfate proteoglycan (CSPG). Has prolyl 3-hydroxylase activity catalyzing the post-translational formation of 3-hydroxyproline in -Xaa-Pro- Gly- sequences in collagens, especially types IV and V. May be involved in the secretory pathway of cells. Has growth suppressive activity in fibroblasts.
Especificaciones
| LEPRE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q32P28 | |
| P3H1 | |
| Full length human recombinant protein of LEPRE1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H3 | |
| Unconjugated | |
| P3H1 | |
| GROS1; Growth suppressor 1; LEPRE1; leprecan; Leprecan-1; leucine- and proline-enriched proteoglycan 1; leucine proline-enriched proteoglycan (leprecan) 1; OI8; P3H1; procollagen-proline 3-dioxygenase; prolyl 3-hydroxylase 1; PSEC0109 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64175 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido