Learn More
Invitrogen™ LDLR Recombinant Rabbit Monoclonal Antibody (SJ0197)

Descripción
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. For Flow cytometry, cells should be fixed and permeabilized before staining.
Especificaciones
Especificaciones
| Antígeno | LDLR |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot, Western Blot |
| Clasificación | Recombinant Monoclonal |
| Clon | SJ0197 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 |
| génica | LDLR |
| N.º de referencia del gen | P01130 |
| Alias de gen | FH; FHC; Hlb301; I79_005860; LDL receptor; LDLA; LDLCQ2; LDLR; ldl-r; LDLRA; low density lipoprotein receptor; low density lipoprotein receptor protein; low-density lipoprotein receptor; low-density lipoprotein receptor class A domain-containing protein 3 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.