Learn More
LC3C Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16868604 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16868604 150 μL 150 microlitros
16878604 20 μL 20 microlitros
2 options
Código de producto. 16868604
Marca: Proteintech 187261AP150UL
Rabbit Polyclonal Antibody
The antibody is specific to the propeptide of LC3C.
MAP1LC3C, also named as LC3C, belongs to the MAP1 LC3 family. It is the human homolog of yeast Atg8 and is involved in the formation of autophagosomal vacuoles, called autophagosomes. LC3C is produced by the proteolytic cleavage after the conserved C-terminal Gly residue. The antibody is specific to the prepeptide of LC3C.
Especificaciones
Antígeno LC3C
Aplicaciones Immunocytochemistry, Immunofluorescence, Western Blot
Clasificación Polyclonal
Concentración 0.23 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica MAP1LC3C
N.º de referencia del gen Q9BXW4
Alias de gen LC3C, MAP1A/MAP1B LC3 C, MAP1A/MAP1B light chain 3 C, MAP1LC3C
Símbolos de los genes MAP1LC3C
Especie del huésped Rabbit
Inmunógeno Peptide
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 440738
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG