missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lauryldimethylamine-N-oxide (LDAO), LC/MS grade

Código de producto. p-200019983 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
1 g
5 g
Tamaño de la unidad:
1 g
5 g
Código de producto. Cantidad unitSize
30245091 1 g 1 g
30245145 5 g 5 g
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30245091

Marca: Thermo Scientific™ A65503

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Lauryldimethylamine-N-oxide (LDAO) is a zwitterionic surfactant that possesses unique properties for solubilization and stabilization of biomolecules, particularly membrane proteins.

Lauryldimethylamine-N-oxide (LDAO) is a zwitterionic surfactant that possesses unique properties for solubilization and stabilization of biomolecules, particularly membrane proteins. LDAO shows particular utility in native mass spectrometry (MS) to study the intact structure and non-covalent interactions of proteins and protein complexes. Incorporation of LDAO into sample preparation can help maintain the integrity of non-covalent interactions, such as protein-protein or protein-ligand interactions, during the transfer from solution to gas phase.

Features of LDAO include:
• Zwitterionic detergent
• Mildness and biocompatibility
• Enhances the stability of solubilized proteins, including membrane proteins
• High-purity compound with low UV absorptivity
• Native mass spec compatible

Applications
LDAO finds applications in various experimental techniques in addition to native MS, including protein purification, crystallization, and structural biology studies. It is compatible with a wide range of biomolecules and can be employed for both soluble and membrane protein research.

Usage
It is important to optimize the concentration of LDAO to ensure that it provides effective solubilization and stabilization of the biomolecules while avoiding excessive detergent interference in the mass spectrometry analysis. Additionally, the specific conditions and parameters of the native MS experiment, such as the choice of ionization source and mass spectrometer parameters, should be carefully considered and optimized for each specific application.

Properties
CMC: (H2O) ~1–2 mM (0.023 %)
Aggregation number: (H2O) ~76
Purity: ≥99% by HPLC analysis (all amine oxides) with <2% other DAOs
pH: 6–8.5 (1% solution in water)
Solubility: ≥30% (in water at 20°C)
Fluorescence: <10% (0.1 solution in water at 345 nm)
Absorbance of a 1% solution in water: 260 nm: <0.08, 280 nm: <0.06, 340 nm: <0.02

TRUSTED_SUSTAINABILITY

Especificaciones

Color Blanco
Tipo de producto Lauryldimethylamine-N-oxide
CAS 1643-20-5
Fórmula molecular C14H31NO
Formulario Polvo
Para utilizar con (aplicación) LC-MS
Cantidad 1 g
Peso molecular 229.4
Pureza ≥99%

Para uso exclusivo en investigación.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.