Learn More
Gilac™ Large Volume Container
Click to view available options
Capacidad (métrico):
100 L
310 L
500 L
Tamaño de la unidad:
cada uno
Descripción
- Excellent mechanical resistance: made from high density polyethylene for increased shock resistance.
- The durable, thick walls mean no distortion and warping.
- Limited gaps and overlapping, equipped with edges so easy to handle.
Especificaciones
Especificaciones
| Tipo | Storage Tote |
| Material | HDPE |
| Color | Blanco |
| Forma | Rectangulares |
| Capacidad (imperial) | 26.4 gal. |
| Capacidad (métrico) | 100 L |
| Altura (imperial) | 2.2 ft. |
| Altura (métrico) | 665 mm |