missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Lapatinib
Click to view available options
Cantidad:
10 mg
Tamaño de la unidad:
10 miligramos
Descripción
- Lapatinib is a selective dual inhibitor of epidermal growth factor receptor (EGFR, ERBB1) and human epidermal growth factor receptor 2 (HER2, ERBB2), each with an IC50 of about 10 nM (Rusnak et al.) It binds reversibly, but with a very slow off-rate, to the ATP binding pocket in an inactive-like conformation, thus preventing autophosphorylation
- Binding is very potent, with Ki values of 3 and 13 nM to EGFR and HER2 respectively (Wood et al.)
- Induces apoptosis in cells overexpressing EGFR and/or HER2, and inhibits tumor growth in a mouse xenograft model (Xia et al.)
Specifications
Specifications
| CAS | 231277-92-2 |
| Sinónimo | GSK 572016; GW 572016; Tyverb; GW2016 |
| Cantidad | 10 mg |
| Fórmula molecular | C29H26ClFN4O4S |
| Pureza | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction