Descripción
Lamina-associated polypeptide 2 (LAP2), isoforms alpha/beta/gamma is a protein that in humans is encoded by the TMPO gene. LAP2 is an inner nuclear membrane (INM) protein. LAP2 plays a role in the regulation of nuclear architecture by binding lamin B1 and chromosomes. This interaction is regulated by phosphorylation during mitosis.
Especificaciones
Especificaciones
| Antígeno | LAP2 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.13 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | TMPO |
| N.º de referencia del gen | P42166, Q61033, Q62733 |
| Alias de gen | CMD1T, LAP2, LEMD4, PRO0868, Splenin Thymopentin, thymopoietin, Thymopoietin isoform alpha, TMPO, TP, TP alpha, TP5, TPRP isoform alpha |
| Símbolos de los genes | Tmpo |
| Mostrar más |