Learn More
Invitrogen™ LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody, 1 mg
Código de producto. 10083614
Change view
Click to view available options
:
1 mg
Código de producto.
10083614 1 mg
1 options
Código de producto. 10083614
Marca: Invitrogen™ PV5129
The p53 tumor suppressor protein is an essential component of many different signaling pathways.
The p53 tumor suppressor protein is an essential component of many different signaling pathways. The LanthaScreen™ antibody detects the phosphorylation of p53 at Ser15 by a range of kinases including DNA-PK, ATM, and ATR.
For use in combination with the Fluorescein-p53 (Ser15) peptide substrate (Cat. No. PV5132) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Ser/Thr kinases
Clasificación Monoclonal
Conjugado Tb (Terbium)/Lanthanide Chelates
Peso molecular del antígeno 150kDa
Cantidad 1 mg
Estado normativo RUO
Disciplina de investigación Pharma and Biopharma
Primario o secundario Primary
Envase 1 Each
Contenido y almacenamiento Terbium-labeled p53 (pSer15) antibody in HEPES buffered saline.
Store at -20°C.
Método de detección Phosphorylation of p53
Excitación/emisión 340/490, 520 nm
Reactividad Human
Condiciones de envío Dry Ice
Presentación de datos End Point
Reactividad abrv Hu
Gen diana Ser/Thr Kinases
Línea de productos LanthaScreen
Mostrar más Mostrar menos
For Research Use Only. Not for use in diagnostic procedures.