Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K27me3 Antibody
Descripción
- The LanthaScreen™ Tb-anti-Histone H3K27me3 antibody is a key component of the BacMam Histone H3K27me3 Cellullar Assay (A14159)
- When this terbium (Tb)- labeled antibody is paired with baculovirus-mediated (BacMam) expression of a Histone H3-GFP fusion protein, the TR-FRET readout enables sensitive detection of H3 lysine 27 trimethylation
- The resulting high-throughput screening (HTS) compatible assay can be used to identify inhibitors of methyltransferases and demethylases acting on lysine 27 of histone H3
- For research use only
- Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K27me3 Antibody Allows Users to:
- Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
- Conserve precious cell samples with the miniaturizable homogenous assay format
- Improve data quality with the advantages of TR-FRET
Especificaciones
Especificaciones
| Concentración | 3500 nM, 0.5 mg/mL |
| Conjugado | Tb (Terbium)/Lanthanide Chelates |
| Cantidad | 1 mg |
| Contenido y almacenamiento | Store the LanthaScreen™ Tb-anti-histone H3K27me3 Antibody at -20° C. |
| Formulario | Liquid |
| Condiciones de envío | Dry Ice |
| Estado de modificación de destino | Methylated |
For Research Use Only. Not for use in diagnostic procedures.