missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-anti-GFP Antibody

Código de producto. 11330732
Change view
Click to view available options
Cantidad:
1 mg
25 μg
Tamaño de la unidad:
cada uno
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11330732 25 μg cada uno
11340732 1 mg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11330732 Proveedor Invitrogen™ N.º de proveedor A13391

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

LanthaScreen™ Tb anti-GFP Antibody is a terbium-labeled antibody recognizing green fluorescent protein (GFP).

LanthaScreen™ Tb anti-GFP Antibody is a terbium-labeled antibody recognizing green fluorescent protein (GFP). It can be used to determine GFP expression levels in any GFP expressing cell line with greater sensitivity than detecting the fluorescence intensity of the GFP protein alone.

With the LanthaScreen™ Tb anti-GFP Antibody You Can:

  • Amplify GFP signal expression for more sensitive results
  • Track GFP-target protein expression levels
  • Easily build TR-FRET assays from any GFP expressing cell line

Detect Low Level GFP Expression

GFP is a commonly used reporter for many types of research. In some instances, the level of GFP-target protein expression is not high enough to generate a suitably strong fluorescence intensity signal. The LanthaScreen™ Tb anti-GFP Antibody can help overcome this problem through the use of TR-FRET assays. Since TR-FRET is a ratiometric readout and does not rely on the intensity of the signal alone, low levels of GFP expression are not a problem.

Use TR-FRET to Track Protein Expression Levels

Using the LanthaScreen™ Tb anti-GFP Antibody in conjunction with a cell line expressing GFP-target fusion protein can allow development of TR-FRET assays for protein accumulation or degradation. Following titration of a compound that induces or inhibits the expression of your GFP-target protein, LanthaScreen™ Tb anti-GFP can be added to the resultant cell lysates and the TR-FRET ratios compared to determine the relative effect of the compound on the GFP-target protein expression level.

Create New TR-FRET Assays from Any GFP Expressing Cell Line

Because the terbium label on the LanthaScreen™ Tb anti-GFP Antibody can be used as a donor for TR-FRET with GFP, any GFP-expressing cell line can be used with the antibody to build a TR-FRET assay. The antibody will bind to GFP in a cell lysate sample, and a simple ratiometric measurement can determine the differences in GFP expression levels between different samples.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno GFP
Aplicaciones Kinase Assay
Concentración 0.54mg/mL
Conjugado Tb (Terbium)/Lanthanide Chelates
Cantidad 25 μg
Estado normativo RUO
Disciplina de investigación Pharma and Biopharma
Primario o secundario Primary
Envase 1 Each
Contenido y almacenamiento Terbium-labeled anti-GFP antibody in HEPES buffered saline.

Store at -20°C.
Formulario Liquid
Condiciones de envío Dry Ice
Molécula diana GFP
Técnica TR-FRET
Línea de productos LanthaScreen
Mostrar más Mostrar menos

For Research Use Only. Not for use in diagnostic procedures.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.