Learn More
Thermo Scientific™ LanthaScreen™ Eu-anti-DYKDDDDK Antibody
Descripción
LanthaScreen Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag). This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity.
Especificaciones
Especificaciones
| Concentración | 0.25 mg⁄ml |
| Conjugado | Alexa Fluor 647/Europium/Lanthanide Chelates |
| Formulación | Frozen |
| Cantidad | 25 μg |
| Estado normativo | RUO |
| Disciplina de investigación | Target and Lead Identification and Validation |
| Primario o secundario | Primary |
| Envase | 1 Each |
| Contenido y almacenamiento | Europium-labeled anti-DYKDDDDK antibody in HEPES buffered saline. Store at -20°C. |
| Formulario | Frozen |
| Mostrar más |
For Research Use Only. Not for use in diagnostic procedures.