Learn More
Invitrogen™ LanthaScreen™ Elite Tb-anti-HIS Antibody
Descripción
LanthaScreen Elite Terbium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Tb chelates negatively impacts protein activity. The Elite Tb-anti-HIS-tag antibody is a specialized antibody with a broader HIS-tag binding performance than our traditional Tb-anti-HIS-tag antibody.
Especificaciones
Especificaciones
| Aplicaciones | Kinase Assay |
| Concentración | 0.46 mg/mL |
| Conjugado | Tb (Terbium)/FITC |
| Peso molecular del antígeno | 150 kDa |
| Cantidad | 25 μg |
| Estado normativo | RUO |
| Disciplina de investigación | Kinase Biology |
| Primario o secundario | Primary |
| Envase | 1 Each |
| Contenido y almacenamiento | Elite Terbium-labeled anti-HIS antibody in HEPES buffered saline. Store at -20°C. |
| Mostrar más |
For Research Use Only. Not for use in diagnostic procedures.