Learn More
Invitrogen™ LAMTOR5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA596186
Descripción
Immunogen sequence: MEPGAGHLDG HRAGSPSLRQ ALCDGSAVMF SSKERGRCTV INFVPLEAPL RSTPRSRQVT EACGGEGRAV PLGSEPEWSV GGMEATLEQH LEDTMKNPSI VGVLCTDSQG LNLGCRGTLS DEHAGVISVL AQQAAKLTSD PTDIPVVCLE SDNGNIMIQK HDGITVAVHK MAS; Positive Samples: U-251MG, Mouse kidney; Cellular Location: Cytoplasm, Lysosome.
HBXIP, also named as XIP, belongs to the HBXIP family. It is originally identified by its interaction with the C-terminus of hepatitis B virus (HBV) X protein (HBx). HBXIP is a regulator of centrosome dynamics and cytokinesis. HBXIP is able to up-regulate S100A4 though activating STAT4 and inducing DNA methylation of PTEN. HBXIP is a cellular 18kd protein and cytoplasm stain.
Especificaciones
| LAMTOR5 | |
| Polyclonal | |
| Unconjugated | |
| LAMTOR5 | |
| 1110003H18Rik; HBV X-interacting protein; HBV X-interacting protein homolog; HBx-interacting protein; HBX-interacting protein homolog; HBXIP; hepatitis B virus x interacting protein; Hepatitis B virus X-interacting protein; hepatitis B virus x-interacting protein (9.6kD); hepatitis B virus X-interacting protein homolog; LAMTOR5; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 5; late endosomal/lysosomal adaptor, MAPK and MTOR activator 5; MGC71071; ragulator complex protein LAMTOR5; XIP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10542, 295357, 68576 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.07 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43504, Q9D1L9 | |
| LAMTOR5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human LAMTOR5 (NP_0063932). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.