missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LAMP1 (CD107a) Biosimilar Recombinant Monoclonal Antibody

Código de producto. 30284153
Change view
Click to view available options
Cantidad:
1 mg
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
30284153 1 mg -
30284452 100 μg 100 microgramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30284153

Marca: Invitrogen™ MA559006

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Monoclonal Antibody

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term SAR428926.

LAMP1 (CD107a, lysosome-associated membrane protein-1) together with LAMP-2, is a major constituent of lysosomal membrane, 1-2% of total CD107a is found also on the plasma membrane. LAMP1 is a heavily glycosylated membrane protein which contains a putative signal peptide, 18 sites for N-linked glycosylation, a single membrane-spanning segment and a short (11 amino acid) cytosolic tail. The LAMP proteins are involved in lysosome biogenesis and are required for fusion of lysosomes with phagosomes. LAMP1 is a type 1 integral membrane protein that is transported from trans-Golgi network to endosomes and then lysosomes. Upon cell activation, LAMP1 transfer to the plasma membrane is dependent on a carboyxl-terminal tyrosine based motif (YXXI). Perturbation in the spacing between the tyrosine based motif relative to the membrane abolishes lysosome localization of LAMP1, and this mutant protein then cycles between the plasma membrane and the endosome. Cell surface LAMP1 (and LAMP2) have been shown to promote adhesion of human peripheral blood mononuclear cells (PBMC) to vascular endothelium, therefore, they are possibly involved in the adhesion of PBMC to the site of inflammation. Increased LAMP1 immunoreactivity is observed in neurons and glial cells surrounding senile plaques in Alzheimer's Disease (AD) cases, and is localized in medullary epithelial cells, single macrophages and lymphocytes in acute thymic involution. LAMP1 is a good marker of mast cell activation.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno LAMP1 (CD107a) Biosimilar
Aplicaciones ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Clasificación Recombinant Monoclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Especie del huésped Human
Método de purificación Protein A
Cantidad 1 mg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.