missing translation for 'onlineSavingsMsg'
Learn More

Laminin Antibody [Janelia Fluor™ 635], Novus Biologicals™

Código de producto. 30175068
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30175068 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30175068 Proveedor Novus Biologicals (Bio-Techne) N.º de proveedor NB300144JF635

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Laminin Polyclonal antibody specifically detects Laminin in Chinese Hamster,Human,Invertebrate,Mammalia,Mouse,Rabbit,Rat,Sheep samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunohistochemistry Free-Floating,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Laminin
Aplicaciones Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunocytochemistry/Immunofluorescence
Clasificación Polyclonal
Conjugado Janelia Fluor 635
Formulación 50mM Sodium Borate
Alias de gen LAMA, Laminin A chain, laminin subunit alpha-1, laminin, alpha 1, Laminin-1 subunit alpha, Laminin-3 subunit alpha, PTBHS, S-LAM alpha, S-laminin subunit alpha
Especie del huésped Rabbit
Inmunógeno Laminin Antibody was made to Laminin 111 isolated from mouse Engelbreth-Holm-Swarm (EHS) sarcoma cells. [UniProt# P19137]
Método de purificación IgG purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 284217
Especies diana Chinese Hamster, Human, Invertebrate, Mammalia, Mouse, Rabbit, Rat, Sheep
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.