Learn More
Lamin B1 Rabbit anti-Human, Mouse, Rat, Clone: 5H10, Proteintech
Rabbit Recombinant Antibody
Marca: Proteintech 80906-1-RR-100UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Lamin B1 antibody apecifically detetects Lamin B1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ELISA.Especificaciones
Lamin B1 | |
5H10 | |
Unconjugated | |
BC012295 | |
Rabbit | |
Protein A purification | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
LMNB1 | |
Lamin B1 fusion protein Ag3631 | |
100 μL | |
4001 | |
Store at -20°C. Stable for one year after shipment. | |
IgG |
Título del producto
Lamin B1 Rabbit anti-Human, Mouse, Rat, Clone: 5H10, Proteintech > 100 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido