missing translation for 'onlineSavingsMsg'
Learn More

LAMC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Código de producto. p-7209169 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Código de producto. Cantidad unitSize
16879174 20 μL 20 microlitros
16869174 150 μL 150 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16879174

Marca: Proteintech 196981AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Laminins, a family of extracellular matrix glycoproteins, are the major noncollagenous constituent of basement membranes. They have been implicated in a wide variety of biological processes including cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. Laminins are composed of 3 non identical chains: laminin alpha, beta and gamma (formerly A, B1, and B2, respectively) and they form a cruciform structure consisting of 3 short arms, each formed by a different chain, and a long arm composed of all 3 chains. Each laminin chain is a multidomain protein encoded by a distinct gene. Several isoforms of each chain have been described. Different alpha, beta and gamma chain isomers combine to give rise to different heterotrimeric laminin isoforms which are designated by Arabic numerals in the order of their discovery, i.e. alpha1beta1gamma1 heterotrimer is laminin 1. The biological functions of the different chains and trimer molecules are largely unknown, but some of the chains have been shown to differ with respect to their tissue distribution, presumably reflecting diverse functions in vivo. This gene encodes the gamma chain isoform laminin, gamma 2. The gamma 2 chain, formerly thought to be a truncated version of beta chain (B2t), is highly homologous to the gamma 1 chain; however, it lacks domain VI, and domains V, IV and III are shorter. It is expressed in several fetal tissues but differently from gamma 1, and is specifically localized to epithelial cells in skin, lung and kidney. The gamma 2 chain together with alpha 3 and beta 3 chains constitute laminin 5 (earlier known as kalinin), which is an integral part of the anchoring filaments that connect epithelial cells to the underlying basement membrane. The epithelium-specific expression of the gamma 2 chain implied its role as an epithelium attachment molecule, and mutations in this gene have been associated with junctional epidermolysis bullosa, a skin disease characterized by blisters due to disruption of the epidermal-dermal junction. Two transcript variants resulting from alternative splicing of the 3′ terminal exon, and encoding different isoforms of gamma 2 chain, have been described. The two variants are differentially expressed in embryonic tissues, however, the biological significance of the two forms is not known. Transcript variants utilizing alternative polyA_signal have also been noted in literature.

Especificaciones

Antígeno LAMC2
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.41 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica LAMC2
N.º de referencia del gen Q13753, Q61092
Alias de gen B2T, BM600, CSF, CSF 140 kDa subunit, EBR2, EBR2A, Epiligrin subunit gamma, Kalinin subunit gamma, Ladsin 140 kDa subunit, LAMB2T, LAMC2, Laminin 5 subunit gamma, Laminin B2t chain, Laminin subunit gamma 2, laminin, gamma 2, LAMNB2, Nicein subunit gamma
Símbolos de los genes LAMC2
Especie del huésped Rabbit
Inmunógeno Peptide
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16782, 192362, 3918
Especies diana Rat, Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.