missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAIR2 Polyclonal Antibody, Invitrogen™
Descripción
The protein encoded by this gene is a member of the immunoglobulin superfamily. It was identified by its similarity to leukocyte-associated immunoglobulin-like receptor 1, a membrane-bound receptor that modulates innate immune response. The protein encoded by this locus is a soluble receptor that may play roles in both inhibition of collagen-induced platelet aggregation and vessel formation during placental implantation. This gene maps to a region of 19q13.4, termed the leukocyte receptor cluster, which contains 29 genes in the immunoglobulin superfamily. Alternatively spliced transcript variants have been described for this gene.
Especificaciones
Especificaciones
| Antígeno | LAIR2 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | LAIR2 |
| N.º de referencia del gen | Q6ISS4 |
| Alias de gen | CD306; CD306 antigen; LAIR2; LAIR-2; leukocyte associated immunoglobulin like receptor 2; leukocyte-associated Ig-like receptor 2; leukocyte-associated Ig-like receptor-2; leukocyte-associated immunoglobulin-like receptor 2; MGC71634 |
| Símbolos de los genes | LAIR2 |
| Mostrar más |
Título del producto
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?